3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(1H-Pyrazol-3-yl)benzotrifluoride, 3-(4-(Trifluoromethyl)phenyl)-1H-pyrazole
PC9988-1G
PC9988-250MG
APOSPC9988-1GEA
105
EUR
APOSPC9988-1G
APOSPC9988-250MG
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole 97%
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole
Formula:
C₁₀H₇F₃N₂ Melting Pt: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569436 CAS Number: 362601-71-6 |