Fluroxypyr 1-Methylheptyl ester, certified
Supplier: Apollo Scientific
Synonyms: 1-Methylheptyl [(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetate, Fluroxypyr Meptyl
BI1013-1G BI1013-250MG BI1013-500MG BI1013-5G
APOSBI1013-1GEA 231 EUR
APOSBI1013-1G APOSBI1013-250MG APOSBI1013-500MG APOSBI1013-5G
Fluroxypyr 1-Methylheptyl ester, certified
Fluroxypyr 1-Methylheptyl ester
Formula: C₁₅H₂₁Cl₂FN₂O₃
MW: 367.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144318
CAS Number: 81406-37-3