6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)
Supplier: Apollo Scientific
Warning
BI1369C-100MG BI1369C-250MG BI1369C-50MG
APOSBI1369C-100MGEA 92.8 EUR
APOSBI1369C-100MG APOSBI1369C-250MG APOSBI1369C-50MG
6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)
6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)
U.S. Patent No. 5,364,767 (to Research Organics, Inc.)
Formula: C₈H₆ClNO₄P·C₇H₁₀N
MW: 354.73 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00216690
CAS Number: 159954-33-3