Succinimide
Supplier: Apollo Scientific
Warning
Synonyms: Pyrrolidine-2,5-dione
BI224-1KG BI224-500G BI224-2.5KG
APOSBI224-1KGEA 43.5 EUR
APOSBI224-1KG APOSBI224-500G APOSBI224-2.5KG
Succinimide
Succinimide
Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 285…290 °C
Melting Pt: 121…126 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005495
CAS Number: 123-56-8
EINECS: 204-635-6