Lithium dodecyl sulphate ≥98%
Supplier: Apollo Scientific
Danger
Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
BI3331-100G BI3331-25G BI3331-5G
APOSBI3331-100GEA 74.1 EUR
APOSBI3331-100G APOSBI3331-25G APOSBI3331-5G
Lithium dodecyl sulphate ≥98%
Lithium dodecyl sulphate
  • Assay (by titration after hydrolysis) 99+%
  • pH (1%) = 6.0 - 9.0
  • Water (by Karl Fischer) < 1.0%
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: 4.1 III