1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2:5,6-Di-o-isopropylidene-alpha-D-allofuranose
BI3857-25G BI3857-5G
APOSBI3857-25GEA 45.1 EUR
APOSBI3857-25G APOSBI3857-5G
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Formula: C₁₂H₂₀O₆
MW: 260.28 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135634
CAS Number: 2595-05-3