(R)-(-)-2-Amino-1-propanol
Supplier: Apollo Scientific
Danger
BIA0108-100G BIA0108-25G
APOSBIA0108-100GEA 174 EUR
APOSBIA0108-100G APOSBIA0108-25G
(R)-(-)-2-Amino-1-propanol
(R)-(-)-2-Amino-1-propanol
Formula: C₃H₉NO
Boiling Pt: 168…176 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00064413
CAS Number: 35320-23-1
UN: 2735
ADR: 8,III