β-Amanitin Amanita phalloides

Supplier: Apollo Scientific
Danger

Synonyms: beta-Amanitin

BIA0311-1MG BIA0311-5MG
APOSBIA0311-1MGEA 284 EUR
APOSBIA0311-1MG APOSBIA0311-5MG
β-Amanitin Amanita phalloides
β-Amanitin
Storage Temperature: Freezer
CAS Number: 21150-22-1
EINECS: 244-244-8
UN: 3462
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR