L-Arginine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
BIA0865-100G BIA0865-1KG BIA0865-500G
APOSBIA0865-100GEA 21.9 EUR
APOSBIA0865-100G APOSBIA0865-1KG APOSBIA0865-500G
L-Arginine hydrochloride
L-Arginine hydrochloride
USP, pyrogen free
Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 226…230 °C
Density: 1.42 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1