Paracetamol 98%
Supplier: Apollo Scientific
Warning
APOSBIA1214-500GEA 101 EUR
APOSBIA1214-500G APOSBIA1214-1KG APOSBIA1214-250G
Paracetamol 98%
Paracetamol
Formula: C₈H₉NO₂
MW: 151,16 g/mol
Melting Pt: 168…172 °C
Density: 1,293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
EINECS: 203-157-5