Sodium chloride Pure BP, Ph. Eur., JP, USP, pharma grade
Supplier: Apollo Scientific
Synonyms: Saline, NaCl, Sea salt, Halite
BIA1371-1KG BIA1371-5KG
APOSBIA1371-1KGEA 43.5 EUR
APOSBIA1371-1KG APOSBIA1371-5KG
Sodium chloride Pure BP, Ph. Eur., JP, USP, pharma grade
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C
Melting Pt: 801 °C
Density: 2.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599