3',5'-Dimethoxy-4'-hydroxyacetophenone 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 4'-Hydroxy-3',5'-dimethoxyacetophenone, 1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one, Acetosyringone, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone
BIA2203-1G BIA2203-25G BIA2203-5G
APOSBIA2203-1GEA 26.1 EUR
APOSBIA2203-1G APOSBIA2203-25G APOSBIA2203-5G
3',5'-Dimethoxy-4'-hydroxyacetophenone 99%
3',5'-Dimethoxy-4'-hydroxyacetophenone
Formula: C₁₀H₁₂O₄
MW: 196.2
Boiling Pt: 335
Melting Pt: 117…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008748
CAS Number: 2478-38-8
EINECS: 219-610-5