Polyethylene glycol 8,000, BioChemica

Supplier: Apollo Scientific
Warning

APOSBIA2204-500GEA 87.3 EUR
APOSBIA2204-500G
Polyethylene glycol 8,000, BioChemica
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Melting Pt: 55 °C
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR