Agmatine sulphate
Supplier: Apollo Scientific
Warning
Synonyms: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate
BIA4003-5G BIA4003-25G
APOSBIA4003-5GEA 21.9 EUR
APOSBIA4003-5G APOSBIA4003-25G
Agmatine sulphate
Agmatine sulphate
Formula: C₅H₁₄N₄
MW: 228.27 g/mol
Melting Pt: 231…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00013109
CAS Number: 2482-00-0
EINECS: 219-617-3