L-Alanine-4-nitroanilide hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: H-Ala-pNA·HCl, L-Alanine 4-nitroanilide hydrochloride
BIA4005-1G BIA4005-5G
APOSBIA4005-1GEA 65.4 EUR
APOSBIA4005-1G APOSBIA4005-5G
L-Alanine-4-nitroanilide hydrochloride
L-Alanine-4-nitroanilide hydrochloride
Formula: C₉H₁₂ClN₃O₃
MW: 245.663 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039088
CAS Number: 31796-55-1
EINECS: 250-811-0