L(-)-Arabitol
Supplier: Apollo Scientific
Warning
Synonyms: L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol
BIA4017-100G BIA4017-50G BIA4017-25G
APOSBIA4017-100GEA 406 EUR
APOSBIA4017-100G APOSBIA4017-50G APOSBIA4017-25G
L(-)-Arabitol
L(-)-Arabitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Melting Pt: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00064290
CAS Number: 7643-75-6