D-(-)-Mannitol Ph. Eur., BP, USP, JP low in endotoxins
Supplier: Apollo Scientific
|
Warning
|
APOSBIA4831-1KGEA
527
EUR
APOSBIA4831-1KG
D-(-)-Mannitol Ph. Eur., BP, USP, JP low in endotoxins
D-(-)-Mannitol
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182,17 g/mol Melting Pt: 164…169 °C Density: 1,52 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...