Cetrimonium bromide

Supplier: Apollo Scientific

APOSBIA6284-100GEA 61.1 EUR
APOSBIA6284-100G APOSBIA6284-500G
Cetrimonium bromide
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR