β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.5%, Ultrapure
Supplier: Apollo Scientific
Warning
Synonyms: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
BIB2002-250G BIB2002-1KG BIB2002-50G
APOSBIB2002-250GEA 102 EUR
APOSBIB2002-250G APOSBIB2002-1KG APOSBIB2002-50G
β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.5%, Ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES)
Biological buffer - useful range 5.5 - 6.7.
  • Heavy metals: <15ppm
  • pKa @ 20°C: 6.15 + 0.1
Formula: C₆H₁₃NO₄S
MW: 195.24 g/mol
Melting Pt: 277…280 °C (dec.)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006181
CAS Number: 4432-31-9
EINECS: 214-478-5