Bromothymol blue sodium salt
Supplier: Apollo Scientific
Warning
Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate
BIB6017-25G BIB6017-50G
APOSBIB6017-25GEA 87 EUR
APOSBIB6017-25G APOSBIB6017-50G
Bromothymol blue sodium salt
Bromothymol blue sodium salt
Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7