5-Bromo-DL-tryptophan
Supplier: Apollo Scientific
Synonyms:
2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
BIB6018-1G
APOSBIB6018-1GEA
162
EUR
APOSBIB6018-1G
5-Bromo-DL-tryptophan
5-Bromo-DL-tryptophan
Formula:
C₁₁H₁₁BrN₂O₂ MW: 283.12 g/mol Melting Pt: 264 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005648 CAS Number: 6548-09-0 |