3-Aminophthalhydrazide monosodium salt
Supplier: Apollo Scientific
Warning
Synonyms: Sodium 5-amino-1,4-dioxo-3,4-dihydro-1H-phthalazin-2-ide, Luminol sodium salt
BIB6093-1G BIB6093-5G
APOSBIB6093-1GEA 36.4 EUR
APOSBIB6093-1G APOSBIB6093-5G
3-Aminophthalhydrazide monosodium salt
3-Aminophthalhydrazide monosodium salt
Formula: C₈H₆N₃NaO₂
MW: 199.15 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00161491
CAS Number: 20666-12-0