1,4-Diaminobutane free base
Supplier: Apollo Scientific
Danger
Synonyms: Putrescine, 1,4-Butanediamine, Tetramethylendiamine
BIB6265-1KG BIB6265-500G BIB6265-25G BIB6265-100G BIB6265-2.5KG
APOSBIB6265-1KGEA 323 EUR
APOSBIB6265-1KG APOSBIB6265-500G APOSBIB6265-25G APOSBIB6265-100G APOSBIB6265-2.5KG
1,4-Diaminobutane free base
1,4-Diaminobutane
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Melting Pt: 25…28 °C
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008235
CAS Number: 110-60-1
EINECS: 203-782-3
UN: 2928
ADR: 6.1+8,II