L(-)-Glutathione (oxidised form)

Supplier: Apollo Scientific
Warning

APOSBIB6313-5GEA 154 EUR
APOSBIB6313-5G APOSBIB6313-25G
L(-)-Glutathione (oxidised form)
L(-)-Glutathione (oxidised form)
Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612,64 g/mol
Melting Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR