Fmoc-Lys(Me3)-OH chloride
Supplier: Apollo Scientific
Synonyms: Fmoc-Lys(Me)₃-OH chloride, Fmoc-Lys(Me)3-OH Chloride, Fmoc-Lys(Me)3-OH Chloride
BIBA1011-100MG BIBA1011-250MG
APOSBIBA1011-100MGEA 56.7 EUR
APOSBIBA1011-100MG APOSBIBA1011-250MG
Fmoc-Lys(Me3)-OH chloride
Fmoc-Lys(Me3)-OH chloride
Formula: C₂₄H₃₁ClN₂O₄
MW: 446.972 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672333
CAS Number: 201004-29-7