trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥99%
Supplier: Apollo Scientific
Synonyms: trans-4-[(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexanecarboxylic acid, Mal-AMCHC, trans-4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxylic acid
BIBP1059-500MG BIBP1059-1G
APOSBIBP1059-500MGEA 82.8 EUR
APOSBIBP1059-500MG APOSBIBP1059-1G
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥99%
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid
Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Freezer
MDL Number: MFCD11519178
CAS Number: 69907-67-1