N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate ≥98%
Supplier: Apollo Scientific
Synonyms: 4-Azido-2,3,5,6-tetrafluorobenzoic Acid N-Succinimidyl Ester, ATFB-SE
BIBP1071-1G BIBP1071-100MG BIBP1071-250MG
APOSBIBP1071-1GEA 696 EUR
APOSBIBP1071-1G APOSBIBP1071-100MG APOSBIBP1071-250MG
N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate ≥98%
N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate
Formula: C₁₁H₄F₄N₄O₄
MW: 332.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00467437
CAS Number: 126695-58-7