Cytosine
Supplier: Apollo Scientific
Warning
Synonyms: 6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol
BIC0713-500G BIC0713-1KG
APOSBIC0713-500GEA 48.2 EUR
APOSBIC0713-500G APOSBIC0713-1KG
Cytosine
Cytosine
Formula: C₄H₅N₃O
MW: 111.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824