2-(Phosphonooxy)benzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: Fosfosal, 2-Carboxyphenyl Phosphate
BIC1059-1G BIC1059-5G
APOSBIC1059-1GEA 75.4 EUR
APOSBIC1059-1G APOSBIC1059-5G
2-(Phosphonooxy)benzoic acid
2-(Phosphonooxy)benzoic acid
Formula: C₇H₇O₆P
MW: 218.10 g/mol
Melting Pt: 162 °C
Storage Temperature: Freezer
MDL Number: MFCD00042645
CAS Number: 6064-83-1
EINECS: 227-993-5