5(6)-Carboxyfluorescein
Supplier: Apollo Scientific
Warning
Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid
BIC1073-500MG BIC1073-5G
APOSBIC1073-500MGEA 27.7 EUR
APOSBIC1073-500MG APOSBIC1073-5G
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
Melting Pt: 275 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00151081
CAS Number: 72088-94-9
EINECS: 276-331-1