1,6-Anhydro-β-D-glucose 98% min
Supplier: Apollo Scientific
Synonyms: 1,6-Anhydro-β-D-glucopyranose, (1R,2S,3S,4R,5R)-6,8-Dioxabicyclo[3.2.1]octane-2,3,4-triol, 1,6-Anhydro-β-D-glucose, (-)-1,6-Anhydro-β-D-glucopyranose, Levoglucosan, (-)-1,6-Anhydro-ß-D-glucopyranose
BICL4018-25G BICL4018-5G
APOSBICL4018-25GEA 163 EUR
APOSBICL4018-25G APOSBICL4018-5G
1,6-Anhydro-β-D-glucose 98% min
1,6-Anhydro-beta-D-glucose
Formula: C₆H₁₀O₅
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063248
CAS Number: 498-07-7