1,6-Anhydro-β-D-cellobiose 98% min
Supplier: Apollo Scientific
Warning
BICL4019-50MG BICL4019-500MG
APOSBICL4019-50MGEA 94.4 EUR
APOSBICL4019-50MG APOSBICL4019-500MG
1,6-Anhydro-β-D-cellobiose 98% min
1,6-Anhydro-beta-D-cellobiose
Formula: C₁₂H₂₀O₁₀
MW: 324.282 g/mol
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD13182343
CAS Number: 35405-71-1