2-Deoxy-2-fluoro-D-glucose 99% min
Supplier: Apollo Scientific
Warning
BICL4075-100MG BICL4075-25MG BICL4075-50MG
APOSBICL4075-100MGEA 517 EUR
APOSBICL4075-100MG APOSBICL4075-25MG APOSBICL4075-50MG
2-Deoxy-2-fluoro-D-glucose 99% min
2-Deoxy-2-fluoro-D-glucose
Formula: C₆H₁₁FO₅
Melting Pt: 169…172 °C
Storage Temperature: Freezer
MDL Number: MFCD00077527
CAS Number: 29702-43-0