1,6:2,3-Dianhydro-β-D-mannopyranose 98% min
Supplier: Apollo Scientific
Synonyms: 1,6:2,3-Dianhydro-β-D-mannose
BICL4079-1G BICL4079-5G BICL4079-2G
APOSBICL4079-1GEA 420 EUR
APOSBICL4079-1G APOSBICL4079-5G APOSBICL4079-2G
1,6:2,3-Dianhydro-β-D-mannopyranose 98% min
1,6:2,3-Dianhydro-beta-D-mannopyranose
Formula: C₆H₈O₄
MW: 144.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18643031
CAS Number: 3868-03-9