Linamarin 99% min

Supplier: Apollo Scientific
Warning

Synonyms: α-Hydroxyisobutyronitrile β-D-glucopyranoside

BICL4127-100MG BICL4127-25MG BICL4127-50MG
APOSBICL4127-100MGEA 837 EUR
APOSBICL4127-100MG APOSBICL4127-25MG APOSBICL4127-50MG
Linamarin 99% min
Linamarin
Formula: C₁₀H₁₇NO₆
Melting Pt: 142…143 °C
Storage Temperature: Freezer
MDL Number: MFCD00036209
CAS Number: 554-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR