Methyl-2,3-O-isopropylidene-5-O-p-toluenesulphonyl-β-D-ribofuranoside 99% min
Supplier: Apollo Scientific
Synonyms: Methyl 2,3-O-isopropylidene-5-O-tosyl-beta-D-ribofuranoside, ((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methyl 4-methylbenzenesulphonate, Methyl 2,3-O-isopropylidene-5-O-tosyl-D-ribonucleoside
BICL4176-10G BICL4176-2G BICL4176-5G
APOSBICL4176-10GEA 1010 EUR
APOSBICL4176-10G APOSBICL4176-2G APOSBICL4176-5G
Methyl-2,3-O-isopropylidene-5-O-p-toluenesulphonyl-β-D-ribofuranoside 99% min
Methyl-2,3-O-isopropylidene-5-O-p-toluenesulphonyl-beta-D-ribofuranoside
Formula: C₁₆H₂₂O₇S
MW: 358.405 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038411
CAS Number: 4137-56-8