1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 99% min
Supplier: Apollo Scientific
Synonyms: Diacetone-D-glucose, (-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, (3aR,5S,6S,6aR)-5-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol, D-Glucose diacetonide
BICL4268-100G BICL4268-250G BICL4268-50G
APOSBICL4268-100GEA 145 EUR
APOSBICL4268-100G APOSBICL4268-250G APOSBICL4268-50G
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 99% min
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose
Formula: C₁₂H₂₀O₆
MW: 260.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005544
CAS Number: 582-52-5