(2-Hydroxylpropyl)-β-cyclodextrine (DS~4.5)
Supplier: Apollo Scientific
Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
BICY1076-25G BICY1076-100G
APOSBICY1076-25GEA 190 EUR
APOSBICY1076-25G APOSBICY1076-100G
(2-Hydroxylpropyl)-β-cyclodextrine (DS~4.5)
(2-Hydroxylpropyl)-β-cyclodextrine
Formula: C₆₃H₁₁₂O₄₂
Storage Temperature: Ambient
MDL Number: MFCD16621721
CAS Number: 128446-35-5