Doxycycline monohydrate
Supplier: Apollo Scientific
Warning
BID0123-100G BID0123-250G BID0123-500G
APOSBID0123-100GEA 132 EUR
APOSBID0123-100G APOSBID0123-250G APOSBID0123-500G
Doxycycline monohydrate
Doxycycline monohydrate
Formula: C₂₂H₂₄N₂O₈·H₂O
MW: 462.46 g/mol (444.44 anhy)
Melting Pt: 167…168 °C
Storage Temperature: Ambient
MDL Number: MFCD02682958
CAS Number: 17086-28-1
EINECS: 209-271-1