2-Deoxy-2-fluoro-D-glucose
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, FDG
BID1009-100MG BID1009-25MG BID1009-50MG
APOSBID1009-100MGEA 306 EUR
APOSBID1009-100MG APOSBID1009-25MG APOSBID1009-50MG
2-Deoxy-2-fluoro-D-glucose
2-Deoxy-2-fluoro-D-glucose
Formula: C₆H₁₁FO₅
MW: 182.15 g/mol
Melting Pt: 170…176 °C
Storage Temperature: Freezer
MDL Number: MFCD00077527
CAS Number: 29702-43-0