2-Deoxy-D(-)-ribose ≥99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
BID533-100G BID533-25G
APOSBID533-100GEA 88.6 EUR
APOSBID533-100G APOSBID533-25G
2-Deoxy-D(-)-ribose ≥99%
2-Deoxy-D(-)-ribose
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 292
Melting Pt: 76…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925