2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3)
Supplier: Apollo Scientific
Warning
Synonyms: Deoxyguanosine triphosphate trisodium salt, 2'-Deoxyguanosine-5'-triphosphate trisodium salt
BID6721-100MG BID6721-50MG
APOSBID6721-100MGEA 212 EUR
APOSBID6721-100MG APOSBID6721-50MG
2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3)
2'-Deoxyguanosine 5'-triphosphate trisodium salt (dGTP-Na3)
Formula: C₁₀H₁₃N₅Na₃O₁₃P₃
Storage Temperature: Freezer
MDL Number: MFCD00080337
CAS Number: 93919-41-6