Fast green FCF
Supplier: Apollo Scientific
Synonyms: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt
BIF1401-10G BIF1401-25G
APOSBIF1401-10GEA 87 EUR
APOSBIF1401-10G APOSBIF1401-25G
Fast green FCF
Fast Green FCF
C.I. 42053
Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
MW: 808.86 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
Merck Index: 13,03970