BIIB 021 free base ≥99%
Supplier: Apollo Scientific
Warning
Synonyms: BIIB021, 6-Chloro-9-((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)-9H-purin-2-amine
BIFK0011-10MG BIFK0011-25MG BIFK0011-5MG
APOSBIFK0011-10MGEA 139 EUR
APOSBIFK0011-10MG APOSBIFK0011-25MG APOSBIFK0011-5MG
BIIB 021 free base ≥99%
BIIB 021
Formula: C₁₄H₁₅ClN₆O
MW: 318.76 g/mol
Melting Pt: 185…186 °C
Storage Temperature: Freezer
MDL Number: MFCD15528939
CAS Number: 848695-25-0