4-Iodo-3-nitrobenzamide ≥99%
Supplier: Apollo Scientific
Warning
Synonyms: Iniparib
BIFK0012-100MG BIFK0012-200MG BIFK0012-500MG
APOSBIFK0012-100MGEA 71.2 EUR
APOSBIFK0012-100MG APOSBIFK0012-200MG APOSBIFK0012-500MG
4-Iodo-3-nitrobenzamide ≥99%
4-Iodo-3-nitrobenzamide
Formula: C₇H₅IN₂O₃
MW: 292.0307 g/mol
Melting Pt: 147…158 °C
Storage Temperature: Freezer
MDL Number: MFCD11110639
CAS Number: 160003-66-7