Carnosic acid free acid ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid

BIFK0050-10MG BIFK0050-25MG BIFK0050-50MG
APOSBIFK0050-10MGEA 94.4 EUR
APOSBIFK0050-10MG APOSBIFK0050-25MG APOSBIFK0050-50MG
Carnosic acid free acid ≥99%
Carnosic acid
Formula: C₂₀H₂₈O₄
Storage Temperature: Freezer
MDL Number: MFCD02259459
CAS Number: 3650-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR