L(-)-Galactose 99%
Supplier: Apollo Scientific
Warning
BIFS0105-250MG BIFS0105-100MG
APOSBIFS0105-250MGEA 203 EUR
APOSBIFS0105-250MG APOSBIFS0105-100MG
L(-)-Galactose 99%
L(-)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00063833
CAS Number: 15572-79-9