Streptomycin sulphate 250 mg/ml
Supplier: Apollo Scientific
BIG1032-10ML BIG1032-25ML
APOSBIG1032-10MLEA 50.9 EUR
APOSBIG1032-10ML APOSBIG1032-25ML
Streptomycin sulphate 250 mg/ml
Streptomycin sulphate
  • Antibodies
Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
Melting Pt: 250
Storage Temperature: Refrigerator
MDL Number: MFCD00037023
CAS Number: 3810-74-0
Merck Index: 12,08983