L(+)-Isoleucine ≥99.5%
Supplier: Apollo Scientific
Warning
Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
BII0711-100G BII0711-500G
APOSBII0711-100GEA 21.9 EUR
APOSBII0711-100G APOSBII0711-500G
L(+)-Isoleucine ≥99.5%
L(+)-Isoleucine
  • Pyrogen free
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2