Ketotifen fumarate
Supplier: Apollo Scientific
Warning
Synonyms: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate
BIK4000-1G BIK4000-250MG
APOSBIK4000-1GEA 26.1 EUR
APOSBIK4000-1G APOSBIK4000-250MG
Ketotifen fumarate
Ketotifen fumarate
Formula: C₁₉H₁₉NOS
MW: 425.50 g/mol
Melting Pt: 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079394
CAS Number: 34580-14-8
EINECS: 252-100-0